Learn More
Medchemexpress LLC L-Alanine, N-[(1,1-dimethylethoxy)carbonyl]-L-valyl- | 70396-18-8 | MFCD22381612 | 98.0% | 288.34 g/mol | C13H24N2O5 | 10 G
SDP
Supplier:  Medchemexpress LLC HYW03870310G
Encompass_Preferred
(tert-Butoxycarbonyl)-L-valyl-L-alanine is a Boc-protected dipeptide used as an amino acid derivative and building block in peptide synthesis and biochemical research. It serves as a protected intermediate for peptide coupling, method development, and analytical standards.
  • CAS 70396-18-8 and molecular formula C13H24N2O5.
  • High purity suitable for synthesis and analysis.
  • White to off-white solid for easy handling and storage.
  • Available in multiple packaging sizes for scale flexibility.
  • Suitable as a Boc-protected building block in peptide coupling reactions.
Catalog No. 50-003-00387