Learn More
Medchemexpress LLC L-alanine, N-(2'-C-methyl-6-O-methyl-P-1-naphthalenyl-5'-guanylyl)-, 2,2-dimethylpropyl ester | 1234490-83-5 | 99.9% | C30H39N6O9P | 10MG
SDP
Supplier:  Medchemexpress LLC HY1333710MG
Encompass_Preferred
BMS-986094 is a nucleotide analogue inhibitor of hepatitis C virus (HCV) RNA-dependent RNA polymerase provided for laboratory research and preclinical studies. It is supplied as a white to off-white solid with documented analytical data and safety information, and is intended for research use only.
  • High purity (99.9%).
  • Solid, white to off-white physical form.
  • Molecular formula C30H39N6O9P; molecular weight 658.64.
  • Supplied in small milligram pack sizes for research applications.
  • Certificate of analysis and safety data sheet available.
  • For research use only; not for human or veterinary use.
Catalog No. 50-002-58195