Learn More
Medchemexpress LLC L-Alanine, N-[(3α,5β,7a,12a)-3,7,12-trihydroxy-24-oxocholan-24-yl]- | 18416-54-1 | 99.2% | 479.65 | 5 MG

Supplier: Medchemexpress LLC HY1687615MG
N-Cholyl-L-alanine is a bile acid amidate, which is found in human fecal samples.
- Target: endogenous metabolite
- Pathway: metabolic enzyme/protease
- Storage: -20°C, sealed storage, away from moisture; in solvent: -80°C, 6 months; -20°C, 1 month (sealed storage, away from moisture)
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.