Learn More
Medchemexpress LLC L-Alanine, N-acetyl- | 97-69-8 | 131.13 | 1 ML
SDP
Supplier:  Medchemexpress LLC HYW00406610MM/1ML
Encompass_Preferred
Ac-Ala-OH is an endogenous metabolite that has been shown to inhibit APEH. This compound can be utilized in breast cancer research.
  • Endogenous metabolite
  • Inhibits APEH
  • Can be used in breast cancer research
  • Inhibits acylaminoacyl peptide hydrolase (APEH) by 42% and 46%
Catalog No. 50-004-45754