Learn More
Medchemexpress LLC L-alanine, N-acetyl- | 97-69-8 | 131.13 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHYW004066R
Encompass_Preferred
Ac-Ala-OH is an endogenous metabolite and an analytical standard intended for research and analytical applications. It inhibits APEH and can be used in breast cancer research.
  • Endogenous metabolite
  • Inhibits APEH
  • Useful in breast cancer research
  • Analytical standard for Ac-Ala-OH
  • Purity: 98.0%
  • White to off-white solid appearance
Catalog No. 50-004-47849