Learn More
Alkali Scientific L Alanine, USP Grade, 100 Grams
SDP
Supplier:  Alkali Scientific LAL001CP
Encompass_Preferred
L-Alanine, USP Grade, is an essential amino acid used in biochemical and pharmaceutical research. This 100g package is ideal for a variety of applications, including enzyme assays, protein synthesis, and as a building block in the synthesis of other amino acids and peptides. L-Alanine plays a role in glucose metabolism and serves as a source of energy for the body. It is commonly used in studies of protein metabolism, nutritional biochemistry, and in the synthesis of pharmaceuticals. This USP-grade reagent ensures high purity and consistency for accurate and reproducible results in laboratory experiments.
Catalog No. 50-006-59934