Learn More
Alkali Scientific L Alanine, USP Grade, 500 Grams
SDP
Supplier:  Alkali Scientific LAL002CP
Encompass_Preferred
L-Alanine, USP Grade, 500 g, is a non-essential amino acid that plays a significant role in the metabolism of glucose and protein synthesis. It is a key player in the transamination process, which helps in the conversion of amino acids into glucose for energy production. This amino acid is commonly used in the food, pharmaceutical, and cosmetic industries. The 500g USP-grade pack ensures high purity and reliability, making it ideal for clinical and biochemical research. L-Alanine is particularly valuable in studies on glucose metabolism, sports nutrition, and energy production, where it supports metabolic processes and protein balance.
Catalog No. 50-006-59905