Learn More
Cayman Chemical L-Alanosine 10mg
SDP
Supplier:  Cayman Chemical 1954510 MG
Encompass_Preferred
An inhibitor of adenylosuccinate synthase (mean IC50 = 4.8 μM and 10 μM in T-ALL and CAK-1 cells, respectively); antineoplastic activity in MTAP deficient cells; disrupts de novo purine biosynthesis and inhibits cellular metabolism
Catalog No. 50-298-5384