Learn More
Medchemexpress LLC L-Alanyl-(4R)-4-hydroxy-L-proline | 76400-25-4 | 99.4% | 202.21 | 50 MG

Supplier: Medchemexpress LLC HYP533850MG
H-Ala-Hyp-OH is a biologically active peptide intended for research use only.
- High purity: 99.36%
- Solid appearance, white to off-white color
- Molecular formula: C8H14N2O4
- Ships at room temperature (continental US)
- Powder storage: -80°C for 2 years, or -20°C for 1 year
- In-solvent storage: -80°C for 6 months, or -20°C for 1 month
- Soluble in DMSO (100 mg/mL), requiring ultrasonic for dissolution
- Classified as a peptide and dipeptide
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.