Learn More
Medchemexpress LLC L-Alanyl-L-alanyl-L-glutamine | 290312-62-8 | 99.0% | 288.30 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYP46215MG
Encompass_Preferred
L-Alanyl-L-alanyl-L-glutamine is a peptide commonly utilized in various biochemical studies. It is recommended for sealed storage, away from moisture, to maintain its stability. When in powder form, it can be stored at -20°C for up to 2 years or at -80°C for 1 year. If stored in solvent, it maintains stability at -80°C for 6 months, or at -20°C for 1 month, also requiring sealed storage away from moisture.
  • Can be used for various biochemical studies.
  • Requires sealed storage, away from moisture.
  • Stable in powder form at -20°C for 2 years.
  • Stable in powder form at -80°C for 1 year.
  • Stable in solvent at -80°C for 6 months.
  • Stable in solvent at -20°C for 1 month.
  • Soluble in H2O at 50 mg/mL (173.43 mM), may require ultrasonic.
Catalog No. 50-003-99175