Learn More
Cayman Chemical L-AlbIzzIIn 500mg
SDP
Supplier:  Cayman Chemical 36665500 MG
Encompass_Preferred
A non-proteinogenic amino acid and metabolite of 5-aminouracil; inhibits 4-methyleneglutaminase (IC50 = 20 mM for the peanut enzyme)
Catalog No. 50-291-0068