Learn More
Medchemexpress LLC L-Allothreonine | 28954-12-3 | 119.12 | 1 ML

Supplier: Medchemexpress LLC HYW00831510MM/1ML
L-Allothreonine is an endogenous metabolite. This product is suitable for research use only.
- Molecular weight: 119.12
- Purity: 98.0%
- Appearance: White to off-white solid
- Solubility in H2O: 12.5 mg/mL (104.94 mM)
- Storage: -20°C for powder, -80°C for solutions
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.