Learn More
Medchemexpress LLC L-Allothreonine (Standard) | 28954-12-3 | 119.12 | 100 MG
SDP
Supplier:  Medchemexpress LLC HYW008315R100MG
Encompass_Preferred
L-Allothreonine (Standard), also known as H-allo-Thr-OH (Standard), is an analytical and reference standard intended for research and analytical applications. It is an endogenous metabolite commonly used in qualitative, quantitative, and methodological research experiments.
  • Analytical standard for L-Allothreonine
  • Intended for research and analytical applications
  • Endogenous metabolite
  • Used in qualitative research experiments
  • Used in quantitative research experiments
  • Used in methodological research experiments
  • Compatible with HPLC, GC, and MS techniques
Catalog No. 50-004-47702