Learn More
Medchemexpress LLC L-Allothreonine (Standard) | 28954-12-3 | 119.12 | 50 MG
SDP
Supplier:  Medchemexpress LLC HYW008315R50MG
Encompass_Preferred
L-Allothreonine (Standard) is the analytical standard of L-Allothreonine. It is an endogenous metabolite intended for research and analytical applications.
  • Used as a reference standard for assay.
  • Utilized in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS.
Catalog No. 50-004-45516