Learn More
Abcam L-alpha-Phosphatidylinositol-3,4,5-trisphosphate sodium salt, PKC delta, epsilon and eta activator, 100UG
Supplier:  Abcam AB145221100UG
Encompass_Preferred
MW 1204.8 g/mol, Purity >98%. Activates Ca²+-insensitive PKC isotypes δ, ε and η in vitro. Binds to the general receptor for phosphoinositide-1 (GRP1) protein through a plekstrin homology (PH) domain.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5387