Learn More
Medchemexpress LLC L-ANAP hydrochloride | 00-00-0 | 99.9% | 308.76 | C15H17ClN2O3 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY101937B5MG
Encompass_Preferred
L-ANAP hydrochloride is a genetically encodable, polarity-sensitive fluorescent unnatural amino acid used as a site-specific probe to monitor local environment polarity and conformational changes in proteins. It is provided as the hydrochloride salt for research use and is soluble in DMSO with very low water solubility.
  • Genetically encodable fluorescent probe for site-specific labeling.
  • Polarity-sensitive fluorescence reports local environmental changes.
  • High purity suitable for biochemical and biophysical assays.
  • DMSO-soluble formulation for straightforward sample preparation.
  • Supplied as a hydrochloride salt for improved stability and handling.
Catalog No. 50-002-48865