Learn More
Medchemexpress LLC L-ANAP hydrochloride | 00-00-0 | 99.9% | 308.76 g/mol | C15H17ClN2O3 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY101937B10MM 1ML
Encompass_Preferred
L-ANAP hydrochloride is a genetically encodable, polarity-sensitive fluorescent unnatural amino acid supplied as a 10 mM solution (1 mL). It enables site-specific incorporation into proteins to probe local environment, dynamics, and conformational changes, serving as a compact fluorescent probe that complements fluorescent proteins in molecular and cellular studies.
  • Genetically encodable for site-specific incorporation into proteins.
  • Polarity-sensitive fluorescence reports local environmental changes.
  • Small, minimally perturbing fluorescent probe for dynamics studies.
  • High purity suitable for biochemical and biophysical applications.
  • Useful complement to fluorescent proteins in cellular experiments.
Catalog No. 50-002-48486