Learn More
TARGETMOL CHEMICALS INC L-Anserine nitrate 50MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T470450MG
Encompass_Preferred
Also available in 10 mg, 25 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. L-Anserine nitrate (L-Anserine nitrate salt) is a dipeptide found in most animal tissues. In model systems it is a potent Antioxidant and scavener of hydroxyl radicals. The compound inhibits nonenzymatic protein glycation induced by aldose and ketose reducing sugars. It is much less potent than L-carnosine as an inhibitor of protein crosslinking. Purity 99.37%
Catalog No. 50-287-8675