Learn More
Enzo Life Sciences L-AP4 (25mg). CAS: 23052-81-5
SDP
Supplier:  Enzo Life Sciences ALX550026M025
Encompass_Preferred
Selective agonist for group III mGluRs. NMDA agonist. Potent synaptic depressant. Alternative name: L(+)-2-Amino-4-phosphonobutanoic acid. Purity: ≥97% (HPLC). Appearance: White crystalline powder. Solubility: Soluble in dilute aqueous base. Long Term Storage: Ambient.
Catalog No. 50-200-9460