Learn More
Medchemexpress LLC L-AP4 monohydrate | 2247534-79-6 | 99.0% | 201.11 g/mol | C4H12NO6P | 10 MG

Supplier: Medchemexpress LLC HY100781B10MG
L-AP4 monohydrate is a selective agonist of group III metabotropic glutamate receptors used in pharmacological and biochemical research to probe mGlu4, mGlu8, mGlu6, and mGlu7 signaling. It is supplied as a monohydrate solid suitable for in vitro studies.
- Selective group III metabotropic glutamate receptor agonist.
- Reported EC50s: mGlu4 0.13 μM; mGlu8 0.29 μM; mGlu6 1.0 μM; mGlu7 249 μM.
- High purity (99.0%) for research applications.
- Molecular weight 201.11 g/mol; formula C4H12NO6P.
- Soluble in DMSO (~10 mg/mL; may require sonication).
- Recommended storage sealed, away from moisture and light; solids at 4°C; in solution -80°C up to 6 months.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.