Learn More
Apexbio Technology LLC L-AP5 79055-67-7 50mg
SDP
Supplier:  Apexbio Technology LLC B620650
Encompass_Preferred
L-AP5 (CAS 79055-67-7) is a selective antagonist of the N-methyl-D-aspartate (NMDA) receptor a subtype of ionotropic glutamate receptors By competitively inhibiting NMDA receptor activation L-AP5 prevents glutamate-induced calcium influx into neurons thereby modulating excitatory neurotransmission within the central nervous system This compound is widely utilized in neurobiological research to investigate the physiological and pathological roles of NMDA receptor-mediated signaling particularly in studies related to neuroprotection and neuropathological conditions
Catalog No. 50-000-90884