Learn More
Medchemexpress LLC α-L-arabinopyranoside, (3β,16β,23R)-16,23:16,30-diepoxy-20-hydroxydammar-24-en-3-yl | 55466-05-2 | 99.9% | 1045.21 g·mol⁻¹ | C52H84O21 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYN066010MG
Encompass_Preferred
Jujuboside B is a plant-derived saponin used in research as an analytical reference and for in vitro pharmacology. It is notable for its activity against platelet aggregation and is supplied in a high-purity form for laboratory use.
  • Bioactive saponin isolated from Zizyphus jujuba.
  • Inhibits platelet aggregation in vitro.
  • High purity (99.9%) suitable for analytical and research use.
  • Molecular weight 1045.21 g·mol⁻¹ and formula C52H84O21.
  • Supplied as a 10 MG vial for small-scale laboratory experiments.
Catalog No. 50-002-95799