Learn More
Cayman Chemical L-ArgInIn-13C6hydrochlorId 5mg
SDP
Supplier:  Cayman Chemical 90040885 MG
Encompass_Preferred
An internal standard for the quantification of L-arginine by GC- or LC-MS
Catalog No. 50-289-4102