Learn More
Alkali Scientific L Arginine, 100 G
SDP
Supplier:  Alkali Scientific LAR005CP
Encompass_Preferred
L-Arginine, 100 g, is an amino acid that plays an important role in the production of nitric oxide, which helps regulate blood flow and cardiovascular health. Arginine is also involved in protein synthesis, immune function, and the detoxification of ammonia from the body. This 100g pack of L-Arginine is highly pure, making it ideal for use in scientific research, dietary supplements, and pharmaceutical formulations. L-Arginine is widely studied for its effects on cardiovascular health, muscle recovery, and immune function, making it a valuable amino acid for both therapeutic and performance-enhancing applications.
Catalog No. 50-006-59900