Learn More
Sigma Aldrich Fine Chemicals Biosciences L-Arginine, 74-79-3, MFCD00002635, 100 G
Supplier:  Sigma Aldrich Fine Chemicals Biosciences A8094100G
Encompass
Linear Formula: H2NC(=NH)NH(CH2)3CH(NH2)CO2H, Molecular Weight: 174.20, from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 98.5-101.0%, Synonym: (S)-2-Amino-5-guanidinopentanoic acid. L-Arginine has been used: as a Roswell park memorial institute medium (RPMI) media component in the isolation and culture of peripheral blood mononuclear cells (PBMCs), as a RPMI media component for tissue culture, and in DMEM medium for the identification and quantification of phosphorylation sites by stable isotope labeling by amino acids in cell culture (SILAC) and LCMS/MS.
Catalog No. NC1030453