Learn More
Medchemexpress LLC L-arginine-13C,d4 (dihydrochloride) | 1786448-45-0 | 252.14 g/mol | C5^13CH12D4Cl2N4O2 | 1 MG

Supplier: Medchemexpress LLC HYW6540581MG
L-Arginine-13C,d4 (dihydrochloride) is an isotopically labeled L-arginine supplied for scientific research and analytical applications. It contains 13C and four deuterium labels and is provided as the dihydrochloride salt. The material is used as a tracer or internal standard for quantitative analysis by NMR, GC-MS, and LC-MS, and is intended for research use only.
- Isotopically labeled with 13C and d4.
- Provided as the dihydrochloride salt.
- Suitable as a tracer or internal standard for NMR, GC-MS, and LC-MS.
- Molecular weight 252.14 g/mol.
- Available in small analytical quantities (1 mg, 5 mg, 10 mg).
- Purity and storage information provided in the Certificate of Analysis.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.