Learn More
Medchemexpress LLC L-Arginine-13C hydrochloride | 94740-43-9 | 211.65 | 50 MG
SDP
Supplier:  Medchemexpress LLC HYN0455AS750MG
Encompass_Preferred
L-Arginine-13C hydrochloride is the 13C-labeled L-Arginine hydrochloride. It acts as a nitrogen donor for the synthesis of nitric oxide, a potent vasodilator known to be deficient during sickle cell crises.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-003-89811