Learn More
Medchemexpress LLC L-arginine-13C6,15N4,d7 hydrochloride | 2483829-29-2 | 10 MG

Supplier: Medchemexpress LLC HYN0455AS410MG
L-Arginine-13C6,15N4,d7 hydrochloride is a stable isotope-labeled amino acid supplied as the hydrochloride salt for use as heavy tracers and internal standards in mass spectrometry, SILAC, and metabolic flux studies.
- Isotopically labeled: 13C6, 15N4, d7.
- Hydrochloride salt form for improved handling and solubility.
- Molecular weight approximately 227.63 g/mol.
- Supplied in small research quantities (10 MG).
- Intended for use as an internal standard in LC-MS/MS and SILAC applications.
- CAS number 2483829-29-2 for unambiguous identification.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.