Learn More
Medchemexpress LLC L-Arginine-13C6,15N4 hydrochloride | 202468-25-5 | 220.59 | 50 MG
SDP
Supplier:  Medchemexpress LLC HYN0455AS850MG
Encompass_Preferred
L-Arginine-13C6,15N4 (hydrochloride) is the 13C- and 15N-labeled L-Arginine hydrochloride. L-Arginine hydrochloride ((S)-(+)-Arginine hydrochloride) is the nitrogen donor for synthesis of nitric oxide, a potent vasodilator that is deficient during times of sickle cell crisis. This compound can be used as a tracer or as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Nitrogen donor for synthesis of nitric oxide
Catalog No. 50-003-89810