Learn More
Selleck Chemical LLC L-Arginine E4831-1G
SDP
Supplier:  Selleck Chemical LLC E48311G
Encompass_Preferred
L-Arginine (L-glutamate) (Arginine glutamate) is a mixture of amino acids that induces apoptosis via hypertonic stress It also promotes gastric relaxation by providing L-arginine for nitric oxide production thereby benefiting gastrointestinal motility and conditions associated with vagus nerve dysfunction
Catalog No. 50-000-43315