Learn More
Apexbio Technology LLC L-Arginine (hydrochloride),500mg CAS# 1119-34-2
SDP
Supplier:  Apexbio Technology LLC C6436500
Encompass_Preferred
L-Arginine (hydrochloride) is the biological precursor of NO which serves as an important signal and effector molecule in animals. L-Arginine is a substrate for NO synthase which catalyzes the oxidation of L-arginine to NO and L-citrulline. NO released by vascular endothelial cells results in relaxation of strips of vascular tissue, as well as inhibition of platelet aggregation and platelet adhesion. L-arginine can be utilized by vascular endothelial cells for NO synthesis, and thereby serves as a potent vasodilator.  Other sizes are also available. Please inqury us for quote.
Catalog No. 50-224-0427