Learn More
AJINOMOTO USA INC L-Arginine HCl 1Kg pail
Supplier:  AJINOMOTO USA INC 32576
Encompass
Chemical Name (2S)-2-Amino-5-guanidinopentanoic Acid Monohydrochloride; Chemical Formula C6H14N4O2·HCl; Molecular Weight 210.66; Nitrogen Content 26.60%; CAS Number 1119-34-2; Assay 98.5 to 101.0%; Description White crystals or crystalline powder 1Kg pail
Catalog No. NC1183333