Learn More
Medchemexpress LLC L-arginine, N-benzoyl-L-alanyl- (TFA) | 71448-11-8 | MFCD13906325 | 99.8% | 463.41 | 1 MG
SDP
Supplier:  Medchemexpress LLC HYP4522A1MG
Encompass_Preferred
Bz-Ala-Arg TFA is a dipeptide and a substrate of human pancreatic carboxypeptidase B and plasma carboxypeptidase N. This product is designed for research use only and has not been fully validated for medical applications.
  • Substrate of human pancreatic carboxypeptidase B
  • Substrate of plasma carboxypeptidase N
  • Biochemical assay reagent
  • Sealed storage, away from moisture and light is recommended
Catalog No. 50-004-00065