Learn More
Medchemexpress LLC L-Arginine (standard) | 74-79-3 | 174.20 | 50 MG
SDP
Supplier:  Medchemexpress LLC HYN0455R50MG
Encompass_Preferred
L-Arginine (Standard) is the analytical standard of L-Arginine. This product is intended for research and analytical applications. It is the substrate for endothelial nitric oxide synthase (eNOS) to generate NO. It is transported into vascular smooth muscle cells by the cationic amino acid transporter family of proteins, where it is metabolized to nitric oxide (NO), polyamines, or L-proline. L-Arginine is a potent vasodilator and can be used to induce experimental acute pancreatitis.
  • Grade of analytical standard.
  • Reference standard supplied assay.
  • Commonly used in qualitative research experiments.
  • Commonly used in quantitative research experiments.
  • Commonly used in methodological research experiments in HPLC, GC, and MS.
Catalog No. 50-003-89996