Learn More
AJINOMOTO USA INC L-Arginine (L-Arg) 1Kg
Supplier:  AJINOMOTO USA INC 325741KG
Encompass
Chemical Name: (2S)-2-Amino-5-guanidinopentanoic Acid; Chemical Formula: C6H14N4O2; Molecular Weight: 174.2000; Nitrogen Content: 0.32160000; CAS Number: 74-79-3; Assay: 98.5 to 101.0%; Description: White crystals or crystalline powder 1kg
Catalog No. NC0664674