Learn More
Research Products International Corp L-Arginine, 100 Grams
SDP
Supplier:  Research Products International Corp A50010100.0
Encompass_Preferred
L-Arginine is one of the 20 amino acids that constitute the building blocks of proteins in living organisms. It is classified as a semi-essential amino acid.
L-Arginine is a common component in laboratory research, particularly in studies related to cell biology, biochemistry, and molecular biology. Its diverse roles make it valuable for understanding various cellular processes.
Catalog No. 50-213-143