Learn More
Cayman Chemical L-ArgInInhydrochlorIde 50g
SDP
Supplier:  Cayman Chemical 2370350 G
Encompass_Preferred
An amino acid and precursor of NO; a substrate for NO synthase; enhances NO release in porcine aortic endothelial cells; induces vasodilation, increases blood flow, and reduces infarct size by 35 and 28% in normotensive and spontaneously hypertensive rats, respectively
Catalog No. 50-299-4535