Learn More
Sigma Aldrich Fine Chemicals Biosciences L-Ascorbic acid 2-phosphate sesquimagnesium salt hydrate, 113170-55-1 (anhydrous), MFCD08063372, 5 g
Supplier:  Sigma Aldrich Fine Chemicals Biosciences A89605G
Encompass
Empirical Formula (Hill Notation): C6H6Mg1.5O9P · xH2O, Molecular Weight: 289.54 (anhydrous basis), ≥95%. L-Ascorbic acid 2-phosphate sesquimagnesium salt hydrate has been used in mesenchymal stem cells (MSC) expansion medium to incubate scaffolds, in Dulbecco′s modified eagle medium (DMEM) for culturing human umbilical cord cells and mesenchymal stem cells.
Catalog No. NC0602549