Learn More
Medchemexpress LLC L-Ascorbic-2-13C acid | 178101-89-8 | >99.9% | 1 MG
SDP
Supplier:  Medchemexpress LLC HYB0166S21MG
Encompass_Preferred
L-Ascorbic acid-13C-1 is the 13C labeled L-Ascorbic acid. L-Ascorbic acid (L-Ascorbate), an electron donor, is an endogenous antioxidant agent. It selectively inhibits Cav3.2 channels with an IC50 of 6.5 μM and is also a collagen. Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process. Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • This compound can be used as a tracer.
  • This compound can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-06311