Learn More
Medchemexpress LLC HY-P1923 1mg Medchemexpress, L-Asparaginase CAS:9015-68-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP19231MG
Encompass_Preferred
Medchemexpress, HY-P1923 1mg L-Asparaginase CAS:9015-68-3 L-Asparaginase (L-​ASNase), a hydrolase that catalyzes the conversion of L-asparagine, used in acute lymphoblastic leukemia treatment. L-Asparaginase depletes L-asparagine from plasma resulting in inhibition of RNA and DNA synthesis with the subsequent blastic cell apoptosis [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4250