Learn More
Medchemexpress LLC L-asparagine-13C4,15N2,d3 monohydrate | 2483829-30-5 | 99.9% | 159.11 | 13C4H7D3 15N2O4 | 100 MG
SDP
Supplier:  Medchemexpress LLC HYW017443S2100MG
Encompass_Preferred
L-Asparagine-13C4,15N2,d3 monohydrate is a stable isotope-labeled form of the amino acid L-asparagine supplied as the monohydrate for research use. It is designed for applications requiring isotopic enrichment, including metabolic tracing, quantitative mass spectrometry, and NMR, where labeled analytes are distinguished from endogenous compounds.
  • Stable isotope labeling (13C4, 15N2, d3) for unambiguous tracing.
  • High chemical purity (99.90%) for consistent analytical results.
  • Monohydrate form with defined molecular weight for accurate calculations.
  • Suitable as an internal standard in mass spectrometry and quantitative assays.
  • Available in small pack sizes for research-scale experiments.
Catalog No. 50-003-00409