Learn More
Medchemexpress LLC L-asparagine-13C4,15N2 monohydrate | 202406-87-9 | 98.0% | 156.0906 g/mol | C4H10N2O4 | 1 MG
SDP
Supplier:  Medchemexpress LLC HYW017443S1MG
Encompass_Preferred
L-asparagine-13C4,15N2 monohydrate is an isotopically labeled amino acid supplied as a monohydrate for use as an internal standard and tracer in analytical chemistry and metabolic studies. It provides a defined mass shift for accurate quantitation and pathway tracing in mass spectrometry workflows.
  • Isotopically labeled for metabolic tracing and mass spectrometry.
  • Monohydrate solid form for easy handling and dissolution.
  • High purity (98.0%) suitable for analytical standards.
  • Available in small milligram quantities for method development.
  • Stable under recommended storage; stock solutions stable at -80°C for extended periods.
Catalog No. 50-003-01639