Learn More
Medchemexpress LLC L-Asparagine-15N2 hydrate | 287484-32-6 | 152.12 | 50 MG
SDP
Supplier:  Medchemexpress LLC HYN0667S250MG
Encompass_Preferred
L-Asparagine-15N2 monohydrate is the 15N-labeled L-Asparagine monohydrate. It is an essential amino acid for leukemic cells and a substrate for L-Asparaginase, a potent anti-leukemic enzyme. L-Asparaginase promotes asparagine and glutamine depletion, inhibiting protein biosynthesis in lymphoblasts, leading to inhibition of RNA and DNA synthesis and subsequent apoptosis. This compound can be used as a tracer or as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis
  • Suitable for NMR, GC-MS, or LC-MS applications
Catalog No. 50-003-88999