Learn More
Medchemexpress LLC L-Asparagine | 70-47-3 | 132.12 | 500 MG
SDP
Supplier:  Medchemexpress LLC HYN0667500MG
Encompass_Preferred
L-Asparagine is an essential amino acid for leukemic cells and a substrate for L-Asparaginase. This potent anti-leukemic enzyme promotes asparagine and glutamine depletion, inhibiting protein biosynthesis in lymphoblasts. The removal of L-asparagine from plasma leads to the inhibition of RNA and DNA synthesis and subsequent apoptosis.
  • Essential amino acid
  • Substrate for L-Asparaginase
  • Inhibits protein biosynthesis in lymphoblasts
  • Promotes RNA and DNA synthesis inhibition
  • Induces apoptosis
  • Biomarker for childhood acute lymphoblastic leukemia
  • Freeze-drying protective agent
  • Solubilizing agent
Catalog No. 50-003-88522