Learn More
Medchemexpress LLC L-aspartic acid (1,4-13C2) | 101247-29-4 | MFCD00144563 | 99.0% | 10 MG
SDP
Supplier:  Medchemexpress LLC HYN0666S610MG
Encompass_Preferred
L-Aspartic acid-1,4-13C2 is a stable-isotope labeled form of L-aspartic acid used as a tracer and internal standard for quantitative analysis by NMR, GC-MS, and LC-MS. It is used in metabolic and pharmacokinetic studies and as a reagent for prodrug preparation targeting colon and cecal tissues.
  • Stable 13C labeling at positions 1 and 4.
  • High purity suitable for analytical applications (99.0%).
  • White to off-white solid with consistent appearance.
  • Applicable as a tracer and internal standard for NMR, GC-MS, and LC-MS quantitation.
  • Defined storage recommendations: 4°C sealed; in solvent: -80°C for 6 months, -20°C for 1 month.
Catalog No. 50-002-98012