Learn More
Medchemexpress LLC L-Aspartic acid | 10 G
SDP
Supplier:  Medchemexpress LLC HYN066610G
Encompass_Preferred
L-Aspartic acid is an amino acid commonly used for preparing prodrugs to target colon and cecal tissues. It is also utilized in the study of inflammatory conditions and is an endogenous metabolite.
  • Used for preparing prodrugs to target colon and cecal tissues.
  • Utilized in the study of inflammatory conditions.
  • Relevant to neurological disease research.
  • Relevant to inflammation/immunology research.
  • Relevant to cardiovascular disease research.
Catalog No. 50-003-88665