Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC L-aspartic acid-13C | 81201-97-0 | MFCD00083887 | 98.0% | 134.10 g/mol | C4H7NO4 | 10 MG
SDP

Supplier:  Medchemexpress LLC HYN0666S10MG

Encompass_Preferred

L-Aspartic acid-13C is an isotopically labeled form of the endogenous amino acid L-aspartic acid provided as a solid reagent for use as a tracer or internal standard in metabolic studies and analytical applications. It offers high purity and compatibility with common analytical solvents.

  • Suitable for metabolic tracing and isotope-labeling experiments.
  • High purity, 98.0%.
  • Molecular weight 134.10 g/mol.
  • Soluble in water and DMSO.
  • Provides reliable performance in mass spectrometry workflows.

Catalog No. 50-002-95911


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.