Learn More
Medchemexpress LLC L-Aspartic acid-13C4,15N | 202468-27-7 | 138.07 | 1 MG
SDP
Supplier:  Medchemexpress LLC HYN0666S11MG
Encompass_Preferred
L-Aspartic acid-13C4,15N is the 13C-labeled and 15N-labeled L-Aspartic acid. L-Aspartic acid is an amino acid that can penetrate the blood-brain barrier. It is commonly used for preparing prodrugs to target colon and cecal tissues and in the study of inflammatory conditions.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-003-88686