Learn More
Medchemexpress LLC L-Aspartic acid-15N | 3715-16-0 | 134.10 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHYN0666S3
Encompass_Preferred
L-Aspartic acid-15N is the 15N-labeled L-Aspartic acid, an amino acid that can penetrate the blood-brain barrier. It is commonly used for preparing prodrugs to target colon and cecal tissues and in the study of inflammatory conditions.
  • Used as a tracer.
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Used for preparing prodrugs to target colon and cecal tissues.
  • Used in the study of inflammatory conditions.
Catalog No. 50-003-89320