Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC L-aspartic acid, 3-hydroxy-, (3S)- | 7298-99-9 | MFCD00673765 | 98.0% | 149.10 g·mol⁻¹ | C4H7NO5 | 1 MG
SDP

Supplier:  Medchemexpress LLC HY1008011MG

Encompass_Preferred

L-threo-3-Hydroxyaspartic acid is a small-molecule research reagent used to inhibit excitatory amino acid transporters (EAATs) in cellular assays. It has been characterized in HEK293 cell lines with reported inhibition constants, and is used in studies of glutamate transport and transporter pharmacology.

  • Potent EAAT inhibitor with Kᵢs of 11 μM (EAAT1), 19 μM (EAAT2), and 14 μM (EAAT3).
  • Suitable for transporter pharmacology and glutamate uptake studies.
  • Molecular formula C4H7NO5 and molecular weight 149.10 g·mol⁻¹.
  • CAS number 7298-99-9 for unambiguous chemical identification.
  • Available in small 1 mg quantities for research applications.
  • Reported purity approximately 98% (supplier listings).

Catalog No. 50-002-12173


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.